Learn More
Medchemexpress LLC Uridine, 2'-deoxy-2'-fluoro- | 784-71-4 | 99.9% | 246.19 | 500 MG
SDP
Catalog No. 5000454375
Encompass_Preferred
2'-Deoxy-2'-fluorouridine is a derivative of the pyrimidine nucleoside uridine. It functions as a nucleoside analog that inhibits the replication of wild-type viruses by binding to the viral RNA. This compound can be incorporated into DNA and RNA in rat and woodchuck models, making it suitable for anti-viral research.
  • Inhibits wild-type virus replication
  • Binds to viral RNA
  • Used in synthesis of RSV polymerase inhibitors
  • Incorporates into DNA and RNA in rat and woodchuck models
  • Suitable for anti-viral research
Catalog No. 50-004-54375 Supplier Medchemexpress LLC Supplier No. HYW013403500MG
2'-Deoxy-2'-fluorouridine is a derivative of the pyrimidine nucleoside uridine. It functions as a nucleoside analog that inhibits the replication of wild-type viruses by binding to the viral RNA. This compound can be incorporated into DNA and RNA in rat and woodchuck models, making it suitable for anti-viral research.
  • Inhibits wild-type virus replication
  • Binds to viral RNA
  • Used in synthesis of RSV polymerase inhibitors
  • Incorporates into DNA and RNA in rat and woodchuck models
  • Suitable for anti-viral research