Learn More
Medchemexpress LLC Uridine, 2'-deoxy-4-thio- | 5580-20-1 | MFCD00058457 | 99.8% | 244.27 g·mol⁻¹ | C9H12N2O4S | 5 MG
SDP
Catalog No. 5000281529
Encompass_Preferred
4-Thio-2'-deoxyuridine is a sulfur-substituted deoxyuridine nucleoside analog supplied as a light-yellow solid and used as a research reagent in studies of DNA synthesis, nucleoside metabolism, and antitumor mechanisms. It is characterized for biochemical and cell-based experiments and provided with supporting analytical data.
  • High purity (99.8%) for research applications.
  • Sulfur-substituted nucleoside for incorporation and metabolism studies.
  • Stable solid form with recommended low-temperature storage.
  • Well-characterized molecular properties (C9H12N2O4S; 244.27 g·mol⁻¹).
  • Available in small quantities suitable for laboratory-scale experiments.
Catalog No. 50-002-81529 Supplier Medchemexpress LLC Supplier No. HYW2517815MG
4-Thio-2'-deoxyuridine is a sulfur-substituted deoxyuridine nucleoside analog supplied as a light-yellow solid and used as a research reagent in studies of DNA synthesis, nucleoside metabolism, and antitumor mechanisms. It is characterized for biochemical and cell-based experiments and provided with supporting analytical data.
  • High purity (99.8%) for research applications.
  • Sulfur-substituted nucleoside for incorporation and metabolism studies.
  • Stable solid form with recommended low-temperature storage.
  • Well-characterized molecular properties (C9H12N2O4S; 244.27 g·mol⁻¹).
  • Available in small quantities suitable for laboratory-scale experiments.