Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Uridine 3',5'-diphosphate | 2922-95-4 | C9H14N2O12P2 | 25 MG
SDP

Catalog No. 5000375076
Encompass_Preferred

Uridine 3′,5′-diphosphate (3′,5′-UDP) is identified as a competitive RNase inhibitor. This compound is suitable for various laboratory research applications.

  • Competitive RNase inhibitor
  • High purity (99.65%)
  • Solid, white to off-white appearance
  • Powder stable for 3 years at -20°C
  • In solvent stable for 6 months at -80°C and 1 month at -20°C
  • Soluble in water at 125 mg/mL (requires sonication)

Catalog No. 50-003-75076 Supplier Medchemexpress LLC Supplier No. 25MGHY154828
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Uridine 3′,5′-diphosphate (3′,5′-UDP) is identified as a competitive RNase inhibitor. This compound is suitable for various laboratory research applications.

  • Competitive RNase inhibitor
  • High purity (99.65%)
  • Solid, white to off-white appearance
  • Powder stable for 3 years at -20°C
  • In solvent stable for 6 months at -80°C and 1 month at -20°C
  • Soluble in water at 125 mg/mL (requires sonication)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.