Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Uridine, 3'-azido-2',3'-dideoxy- | 84472-85-5 | 99.1% | 253.21 g·mol⁻¹ | C9H11N5O4 | 10 MG
SDP

Catalog No. 5000240753
Encompass_Preferred

3′-Azido-2′,3′-dideoxyuridine is an azide-functionalized nucleoside analogue used as an inhibitor of HIV replication and as a reagent for bioorthogonal click chemistry. It is supplied as a white to off-white solid for research use with defined purity and storage recommendations.

  • Contains an azide group enabling copper-catalyzed and strain-promoted azide-alkyne cycloadditions.
  • High purity (99.11%).
  • Molecular weight 253.21 g·mol⁻¹.
  • Supplied as a 10 mg solid vial suitable for small-scale experiments.
  • Storage: powder -20°C (stable for 3 years); in solution -80°C (6 months) or -20°C (1 month).
  • Applications include antiviral research and bioconjugation studies.

Catalog No. 50-002-40753 Supplier Medchemexpress LLC Supplier No. HY10685010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3′-Azido-2′,3′-dideoxyuridine is an azide-functionalized nucleoside analogue used as an inhibitor of HIV replication and as a reagent for bioorthogonal click chemistry. It is supplied as a white to off-white solid for research use with defined purity and storage recommendations.

  • Contains an azide group enabling copper-catalyzed and strain-promoted azide-alkyne cycloadditions.
  • High purity (99.11%).
  • Molecular weight 253.21 g·mol⁻¹.
  • Supplied as a 10 mg solid vial suitable for small-scale experiments.
  • Storage: powder -20°C (stable for 3 years); in solution -80°C (6 months) or -20°C (1 month).
  • Applications include antiviral research and bioconjugation studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.