Learn More
Medchemexpress LLC Uridine, 3'-deoxy-3'-fluoro- | 57944-13-5 | C9H11FN2O5 | 5 MG
SDP
Catalog No. 5000373667
Encompass_Preferred
3'-Deoxy-3'-fluorouridine is a purine nucleoside analog with a molecular formula of C9H11FN2O5. It appears as a white to off-white solid and has a high purity of 99.66% as determined by LCMS. This product is intended for research use only and has not been fully validated for medical applications.
  • White to off-white solid appearance
  • Purity of 99.66% (LCMS)
  • Consistent with structure based on 1H NMR spectrum and LCMS
Catalog No. 50-003-73667 Supplier Medchemexpress LLC Supplier No. HY1527655MG
3'-Deoxy-3'-fluorouridine is a purine nucleoside analog with a molecular formula of C9H11FN2O5. It appears as a white to off-white solid and has a high purity of 99.66% as determined by LCMS. This product is intended for research use only and has not been fully validated for medical applications.
  • White to off-white solid appearance
  • Purity of 99.66% (LCMS)
  • Consistent with structure based on 1H NMR spectrum and LCMS