Learn More
Medchemexpress LLC Uridine,4-thio- | 13957-31-8 | 99.98% | 260.27 | 1 ML
SDP
Catalog No. 5000454064
Encompass_Preferred
4-Thiouridine is a ribonucleoside analog widely used in RNA analysis and (m)RNA labeling. It works by inhibiting rRNA synthesis, which leads to a nucleolar stress response. It is intended for research use only and is not sold to patients.
  • Ribonucleoside analog
  • Used in RNA analysis and mRNA labeling
  • Inhibits rRNA synthesis
  • Causes a nucleolar stress response
  • High purity: 99.98%
  • Labels and purifies nascent rRNA at 0-10 μM
  • Strongly inhibits rRNA production and processing at ≥50 μM
Catalog No. 50-004-54064 Supplier Medchemexpress LLC Supplier No. HYW01179310MM/1ML
4-Thiouridine is a ribonucleoside analog widely used in RNA analysis and (m)RNA labeling. It works by inhibiting rRNA synthesis, which leads to a nucleolar stress response. It is intended for research use only and is not sold to patients.
  • Ribonucleoside analog
  • Used in RNA analysis and mRNA labeling
  • Inhibits rRNA synthesis
  • Causes a nucleolar stress response
  • High purity: 99.98%
  • Labels and purifies nascent rRNA at 0-10 μM
  • Strongly inhibits rRNA production and processing at ≥50 μM