Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

URIDINE-5'-DIPHOSPHATE, MP Biomedicals

Catalog No. ICN10120525 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
500 mg
1 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10120525 25 mg
ICN10120580 100 mg
ICN10120590 500 mg
ICN10120591 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN10120525 Supplier MP Biomedicals, Inc Supplier No. 0210120525
Add to Cart
Add to Cart

Disodium Salt

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58-98-0
Molecular Formula C9H14N2O12P2
Molecular Weight (g/mol) 404.16
InChI Key XCCTYIAWTASOJW-UHFFFAOYNA-N
Synonym uridine 5'-diphosphate, uridine diphosphate, uridine 5'-trihydrogen diphosphate, udp, uridine-5'-diphosphate, unii-5g0f599a1y, 5'-udp, uridine 5'-pyrophosphate, uridine 5'-pyrophosphoric acid, uridine-diphosphate
PubChem CID 6031
ChEBI CHEBI:17659

Specifications

Quantity 25 mg
Formula Weight 448.17 (Anhydrous)
Chemical Name or Material URIDINE-5'-DIPHOSPHATE, MP Biomedicals
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.