Learn More
Medchemexpress LLC Uridine 5'-diphosphate (sodium salt) | 21931-53-3 | 98.0% | 10 G
SDP
Catalog No. 5000366330
Encompass_Preferred
Uridine 5'-diphosphate sodium salt is a potent, selective P2Y6 receptor native agonist (EC50=300 nM; pEC50=6.52) and a potent P2Y14 antagonist (pEC50=7.28). It is an endogenous metabolite that catalyzes the glucuronidation of a wide array of substrates and is used in nucleic acid (RNA) biosynthesis.
  • Potent, selective P2Y6 receptor native agonist.
  • Potent P2Y14 antagonist.
  • Endogenous metabolite.
  • Catalyzes glucuronidation of various substrates.
  • Used in nucleic acid (RNA) biosynthesis.
  • Induces microglial CCL2 and CCL3 mRNA expression.
  • Induces chemokine expression in microglia and astrocytes through activation of the P2Y6 receptor.
  • Induces activation of NFATc1 and NFATc2 in microglia.
Catalog No. 50-003-66330 Supplier Medchemexpress LLC Supplier No. HYW01082010G
Uridine 5'-diphosphate sodium salt is a potent, selective P2Y6 receptor native agonist (EC50=300 nM; pEC50=6.52) and a potent P2Y14 antagonist (pEC50=7.28). It is an endogenous metabolite that catalyzes the glucuronidation of a wide array of substrates and is used in nucleic acid (RNA) biosynthesis.
  • Potent, selective P2Y6 receptor native agonist.
  • Potent P2Y14 antagonist.
  • Endogenous metabolite.
  • Catalyzes glucuronidation of various substrates.
  • Used in nucleic acid (RNA) biosynthesis.
  • Induces microglial CCL2 and CCL3 mRNA expression.
  • Induces chemokine expression in microglia and astrocytes through activation of the P2Y6 receptor.
  • Induces activation of NFATc1 and NFATc2 in microglia.