Learn More
Medchemexpress LLC Uridine 5'-diphosphate (sodium salt) | 21931-53-3 | 98.0% | 500 MG
SDP
Catalog No. 5000451743
Encompass_Preferred
Uridine 5'-diphosphate sodium salt is a potent and selective P2Y6 receptor native agonist (EC50=300 nM; pEC50=6.52) and a potent P2Y14 antagonist (pEC50=7.28). This endogenous metabolite catalyzes the glucuronidation of a wide array of substrates and is utilized in nucleic acid (RNA) biosynthesis.
  • Potent and selective P2Y6 receptor native agonist.
  • Potent P2Y14 antagonist.
  • Catalyzes glucuronidation of various substrates.
  • Utilized in nucleic acid (RNA) biosynthesis.
Catalog No. 50-004-51743 Supplier Medchemexpress LLC Supplier No. HYW010820500MG
Uridine 5'-diphosphate sodium salt is a potent and selective P2Y6 receptor native agonist (EC50=300 nM; pEC50=6.52) and a potent P2Y14 antagonist (pEC50=7.28). This endogenous metabolite catalyzes the glucuronidation of a wide array of substrates and is utilized in nucleic acid (RNA) biosynthesis.
  • Potent and selective P2Y6 receptor native agonist.
  • Potent P2Y14 antagonist.
  • Catalyzes glucuronidation of various substrates.
  • Utilized in nucleic acid (RNA) biosynthesis.