Learn More
Sigma Aldrich Fine Chemicals Biosciences Uridine 5 diphospho N acet25MG
Catalog No. 502914280
Encompass
Uridine diphosphate (UDP)-GalNAc acts as a precursor in the biosynthesis of O-linked oligosaccharide. It also acts as a substrate for synthesis of chitin a vital element of cell walls in fungi and of exoskeletons of arthropods and insects.
Catalog No. 50-291-4280 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. U525225MG
Uridine diphosphate (UDP)-GalNAc acts as a precursor in the biosynthesis of O-linked oligosaccharide. It also acts as a substrate for synthesis of chitin a vital element of cell walls in fungi and of exoskeletons of arthropods and insects.