Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Uridine-5'-diphosphogalactose Disodium Salt, ≥98%, MP Biomedicals™

Catalog No. ICN19590510 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19590510 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19590510 Supplier MP Biomedicals, Inc Supplier No. 0219590510
Add to Cart
Add to Cart

Chemical Identifiers

CAS 137868-52-1
Molecular Formula C15H22N2Na2O17P2
Molecular Weight (g/mol) 610.27
MDL Number MFCD00077895
InChI Key PKJQEQVCYGYYMM-UHFFFAOYNA-L
Synonym udp-a-d-galactose disodium salt
PubChem CID 126969254
IUPAC Name disodium [5-(2,4-dioxo-1,2,3,4-tetrahydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl {[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl phosphonato]oxy}phosphonate
SMILES [Na+].[Na+].OCC1OC(OP([O-])(=O)OP([O-])(=O)OCC2OC(C(O)C2O)N2C=CC(=O)NC2=O)C(O)C(O)C1O

Specifications

Assay Percent Range ≥98%
Quantity 10 mg
Formula Weight 610.3
Physical Form Powder
Chemical Name or Material Uridine-5'-diphosphogalactose Disodium Salt
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.