Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Uridine-5'-diphosphogalactose Disodium Salt, MP Biomedicals™

Catalog No. MP219590505 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219590505 5 mg
MP219590525 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP219590505 Supplier MP Biomedicals, Inc Supplier No. 0219590505
Add to Cart
Add to Cart

Uridine-5'-diphosphogalactose Disodium Salt ≥95%, Powder

  • UDP-Gal and its analogues are used to study the distribution, specificity and kinetics of galactosyltransferase(s).
  • It is a donor substrate for galactosyltransferases involved in the addition of galatose (galactosylation) molecules to N-linked and O-linked oligosaccharides and glycerolipids(biosynthesis of galactose-containing oligosaccharides).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00077895
Synonym UDPG, UDP-Galactose

Specifications

Molecular Formula C15H22N2O17P2Na2
Quantity 5 mg
Solubility Information Soluble in water (100mg/mL-clear colorless solution).
Molecular Weight (g/mol) 610.3
Physical Form Powder
Grade ≥95%
storageNote1 Store at -0°C, desiccate.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.