Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Uridine-5'-Diphosphoglucose Disodium Salt , MP Biomedicals™

Catalog No. MP210120894 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
1 g
5 g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP210120894 5 g
MP210120880 100 mg
MP210120883 250 mg
MP210120890 500 mg
MP210120891 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MP210120894 Supplier MP Biomedicals, Inc Supplier No. 0210120894
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Uridine-5'-diphosphoglucose Disodium Salt is a biosynthetic product produced by the reaction of UTP and glucose-1-phosphate catalyzed by uridyl transferase.

  • Endogenous nucleotide sugar involved in glycosyltransferase reactions in metabolism
  • Shown to bind the P2Y14 receptor, an atypical P2Y receptor involved in the activation of dendritic cells and glial cells
  • Binds to and activates GPR17, inducing oligodendrocyte differentiation at a maximal concentration of 100μM
TRUSTED_SUSTAINABILITY

Specifications

Color White
Packaging Glass
Quantity 5 g
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.