Learn More
Medchemexpress LLC Uridine-5'-diphosphoglucose pyrophosphorylase | 9026-22-6 | 1 KU
SDP
Catalog No. 5000397333
Encompass_Preferred
Uridine-5'-diphosphoglucose pyrophosphorylase is an enzyme involved in the catalysis of UDP-glucose synthesis. It can also be utilized to determine the concentration of pyrophosphate in urine.
  • Enzyme for UDP-glucose synthesis
  • Assists in pyrophosphate determination in urine
  • Related to glycobiology
  • Recombinant enzyme
  • Associated with metabolic disease
  • Metabolic enzyme/protease
  • Endogenous metabolite
Catalog No. 50-003-97333 Supplier Medchemexpress LLC Supplier No. HYP30181KU
Uridine-5'-diphosphoglucose pyrophosphorylase is an enzyme involved in the catalysis of UDP-glucose synthesis. It can also be utilized to determine the concentration of pyrophosphate in urine.
  • Enzyme for UDP-glucose synthesis
  • Assists in pyrophosphate determination in urine
  • Related to glycobiology
  • Recombinant enzyme
  • Associated with metabolic disease
  • Metabolic enzyme/protease
  • Endogenous metabolite