Learn More
Medchemexpress LLC Uridine-5'-diphosphoglucose pyrophosphorylase | 9026-22-6 | MFCD00132561 | 500 U
SDP
Catalog No. 5000398308
Encompass_Preferred
Uridine-5'-diphosphoglucose pyrophosphorylase is an enzyme involved in the catalysis of UDP-glucose synthesis. It can be used to determine the concentration of pyrophosphate in urine.
  • Catalyzes UDP-glucose synthesis
  • Utilized in pyrophosphate concentration determination
  • Classified as an enzyme
  • Glycobiology enzyme
  • Recombinant
  • Metabolic enzyme/protease
Catalog No. 50-003-98308 Supplier Medchemexpress LLC Supplier No. HYP3018500U
Uridine-5'-diphosphoglucose pyrophosphorylase is an enzyme involved in the catalysis of UDP-glucose synthesis. It can be used to determine the concentration of pyrophosphate in urine.
  • Catalyzes UDP-glucose synthesis
  • Utilized in pyrophosphate concentration determination
  • Classified as an enzyme
  • Glycobiology enzyme
  • Recombinant
  • Metabolic enzyme/protease