Learn More
Medchemexpress LLC Uridine 5′-diphosphoglucose (Standard) | 133-89-1 | 99.59% | 566.30 | 50 MG
SDP
Catalog No. 5000439592
Encompass_Preferred
Uridine 5′-diphosphoglucose (Standard) is the analytical standard of Uridine 5′-diphosphoglucose (HY-113044) and is intended for research and analytical applications. It acts as a potent agonist of the proinflammatory P2Y14 receptor, secreted by cardiomyocytes during ischemia and reperfusion, playing an important role in inflammation regulation and neutrophil polarization. It is also a precursor of glucose-containing oligosaccharides, polysaccharides, glycoproteins, and glycolipids in animal tissues and some microorganisms.
  • Suitable for research in counteracting myocardial infarction/reperfusion (MIR)-induced inflammation in heart tissue.
  • Used as an analytical standard in qualitative, quantitative, and methodological research experiments.
  • Applicable in techniques such as HPLC, GC, and MS.
Catalog No. 50-004-39592 Supplier Medchemexpress LLC Supplier No. HY113044R50MG
Uridine 5′-diphosphoglucose (Standard) is the analytical standard of Uridine 5′-diphosphoglucose (HY-113044) and is intended for research and analytical applications. It acts as a potent agonist of the proinflammatory P2Y14 receptor, secreted by cardiomyocytes during ischemia and reperfusion, playing an important role in inflammation regulation and neutrophil polarization. It is also a precursor of glucose-containing oligosaccharides, polysaccharides, glycoproteins, and glycolipids in animal tissues and some microorganisms.
  • Suitable for research in counteracting myocardial infarction/reperfusion (MIR)-induced inflammation in heart tissue.
  • Used as an analytical standard in qualitative, quantitative, and methodological research experiments.
  • Applicable in techniques such as HPLC, GC, and MS.