Learn More
Medchemexpress LLC Uridine 5'-diphosphoglucose (Standard) | 133-89-1 | 99.6% | 566.30 | 25 MG
SDP
Catalog No. 5000439815
Encompass_Preferred
Uridine 5′-diphosphoglucose (Standard) (UDP-D-Glucose (Standard)) is an analytical standard of Uridine 5′-diphosphoglucose. This product is intended for research and analytical applications. It is a potent agonist of the proinflammatory P2Y14 receptor and plays an important role in the regulation of inflammation and neutrophil polarization in neutrophils. It is also a precursor of glucose-containing oligosaccharides, polysaccharides, glycoproteins, and glycolipids in animal tissues and some microorganisms. It is promising for research in counteracting myocardial infarction/reperfusion (MIR)-induced inflammation in heart tissue.
  • The compound is the grade of analytical standard, which is the reference standard supplied assay.
  • Commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.
  • For research use only.
Catalog No. 50-004-39815 Supplier Medchemexpress LLC Supplier No. 25MGHY113044R
Uridine 5′-diphosphoglucose (Standard) (UDP-D-Glucose (Standard)) is an analytical standard of Uridine 5′-diphosphoglucose. This product is intended for research and analytical applications. It is a potent agonist of the proinflammatory P2Y14 receptor and plays an important role in the regulation of inflammation and neutrophil polarization in neutrophils. It is also a precursor of glucose-containing oligosaccharides, polysaccharides, glycoproteins, and glycolipids in animal tissues and some microorganisms. It is promising for research in counteracting myocardial infarction/reperfusion (MIR)-induced inflammation in heart tissue.
  • The compound is the grade of analytical standard, which is the reference standard supplied assay.
  • Commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.
  • For research use only.