Learn More
Medchemexpress LLC HY-W013175 100mg Medchemexpress, Uridine 5'-monophosphate disodium salt CAS:3387-36-8 Purity:>98%
SDP
Catalog No. 501934580
Encompass_Preferred
Medchemexpress, HY-W013175 100mg Uridine 5'-monophosphate disodium salt CAS:3387-36-8 Uridine 5'-monophosphate disodium salt is component used for RNA synthesis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4580 Supplier Medchemexpress LLC Supplier No. HYW013175100MG
Medchemexpress, HY-W013175 100mg Uridine 5'-monophosphate disodium salt CAS:3387-36-8 Uridine 5'-monophosphate disodium salt is component used for RNA synthesis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.