Learn More
Medchemexpress LLC Uridine 5'-monophosphate-13C9,15N2 disodium | 58-97-9 | 99.8% | 379.07 | 1 MG
SDP
Catalog No. 5000454415
Encompass_Preferred
Uridine 5'-monophosphate-13C9,15N2 disodium is the 13C and 15N labeled Uridine 5'-monophosphate disodium salt. Uridine 5'-monophosphate (5'-Uridylic acid) disodium salt is an orally active mitochondrial ATP-dependent potassium channel activator that has a protective effect on the heart. Uridine 5'-monophosphate disodium salt can promote the synthesis of CDP-choline and induce apoptosis in intestinal epithelial cells, which is beneficial for gut development and reduces diarrhea.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-54415 Supplier Medchemexpress LLC Supplier No. HYW013175S21MG
Uridine 5'-monophosphate-13C9,15N2 disodium is the 13C and 15N labeled Uridine 5'-monophosphate disodium salt. Uridine 5'-monophosphate (5'-Uridylic acid) disodium salt is an orally active mitochondrial ATP-dependent potassium channel activator that has a protective effect on the heart. Uridine 5'-monophosphate disodium salt can promote the synthesis of CDP-choline and induce apoptosis in intestinal epithelial cells, which is beneficial for gut development and reduces diarrhea.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.