Learn More
Medchemexpress LLC Uridine 5'-monophosphate-15N2 | MFCD00067346 | 98.0% | 326.17 g/mol | C9H13 15N2O9P | 1 MG
SDP
Catalog No. 5000246615
Encompass_Preferred
Isotopically labeled uridine 5'-monophosphate (15N2) supplied as a high-purity analytical standard for quantitative mass spectrometry, metabolic tracing, and isotopic dilution assays. Provided as a 1 mg vial of 15N2-labeled compound; molecular weight ≈326.17 g/mol and typical purity ~98%.
  • Isotopically labeled for tracer and mass spectrometry applications.
  • High purity suitable for quantitative analytical assays.
  • Small 1 mg vial size for standards and method development.
  • Suitable as an internal standard in isotopic dilution workflows.
  • Useful for metabolic tracing and pathway studies.
Catalog No. 50-002-46615 Supplier Medchemexpress LLC Supplier No. HY101981S1MG
Isotopically labeled uridine 5'-monophosphate (15N2) supplied as a high-purity analytical standard for quantitative mass spectrometry, metabolic tracing, and isotopic dilution assays. Provided as a 1 mg vial of 15N2-labeled compound; molecular weight ≈326.17 g/mol and typical purity ~98%.
  • Isotopically labeled for tracer and mass spectrometry applications.
  • High purity suitable for quantitative analytical assays.
  • Small 1 mg vial size for standards and method development.
  • Suitable as an internal standard in isotopic dilution workflows.
  • Useful for metabolic tracing and pathway studies.