Learn More
Medchemexpress LLC Uridine 5'-monophosphate | 58-97-9 | MFCD00006540 | 324.18 g/mol | C9H13N2O9P | 10 MG
SDP
Catalog No. 5000247249
Encompass_Preferred
Uridine 5'-monophosphate is the monophosphate nucleotide of uridine, supplied as an analytical reference material for research and analytical applications. It is intended for use in biochemical studies, assay calibration, and method development, and is characterized by molecular formula C9H13N2O9P, molecular weight 324.18 g/mol, and CAS 58-97-9.
  • Provided as an analytical reference standard for research use.
  • Supports calibration and quantitation in nucleotide assays.
  • Useful for studies of nucleotide metabolism and enzymology.
  • Soluble in water and DMSO at high concentrations (approximately 150 mg/mL).
  • Supplied in a small pack suitable for analytical testing and method development.
Catalog No. 50-002-47249 Supplier Medchemexpress LLC Supplier No. HY101981R10MG
Uridine 5'-monophosphate is the monophosphate nucleotide of uridine, supplied as an analytical reference material for research and analytical applications. It is intended for use in biochemical studies, assay calibration, and method development, and is characterized by molecular formula C9H13N2O9P, molecular weight 324.18 g/mol, and CAS 58-97-9.
  • Provided as an analytical reference standard for research use.
  • Supports calibration and quantitation in nucleotide assays.
  • Useful for studies of nucleotide metabolism and enzymology.
  • Soluble in water and DMSO at high concentrations (approximately 150 mg/mL).
  • Supplied in a small pack suitable for analytical testing and method development.