Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Uridine 5'-monophosphate | 58-97-9 | MFCD00067346 | 100.0% | 324.18 | C9H13N2O9P | 25 MG
SDP

Catalog No. 5000247980
Encompass_Preferred

Uridine 5'-monophosphate is a ribonucleotide provided as a high-purity reference standard intended for research and analytical applications. It is suitable for use in biochemical assays, analytical method development, and as a calibration standard.

  • High purity 99.98%
  • Molecular formula C9H13N2O9P
  • Molecular weight 324.18 g/mol
  • CAS number 58-97-9
  • Pack size 25 mg
  • Suitable as a reference standard for biochemical assays and analytical method development

Catalog No. 50-002-47980 Supplier Medchemexpress LLC Supplier No. HY101981R25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Uridine 5'-monophosphate is a ribonucleotide provided as a high-purity reference standard intended for research and analytical applications. It is suitable for use in biochemical assays, analytical method development, and as a calibration standard.

  • High purity 99.98%
  • Molecular formula C9H13N2O9P
  • Molecular weight 324.18 g/mol
  • CAS number 58-97-9
  • Pack size 25 mg
  • Suitable as a reference standard for biochemical assays and analytical method development

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.