Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Uridine 5'-monophosphate | 58-97-9 | MFCD00067346 | 100.0% | 324.18 g/mol | C9H13N2O9P | 1 ML
SDP

Catalog No. 5000246614
Encompass_Preferred

Uridine 5'-monophosphate is a ribonucleotide used as a biochemical reagent, analytical reference, and building block in RNA-related assays and metabolic studies. It is supplied as a solid or as ready-to-use solutions for laboratory workflows and quality-control applications.

  • Ribonucleotide reagent for biochemical and cell-culture assays.
  • CAS 58-97-9; molecular formula C9H13N2O9P; molecular weight 324.18 g/mol.
  • Available as powder or prepared solutions (examples: 10 mM solution, 1 mL ampoules).
  • High purity reported (typically ≈99.8-99.98% in supplier documentation).
  • Soluble in water and DMSO; powder storage -20 °C, solutions typically -80 °C for long-term storage.

Catalog No. 50-002-46614 Supplier Medchemexpress LLC Supplier No. HY10198110MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Uridine 5'-monophosphate is a ribonucleotide used as a biochemical reagent, analytical reference, and building block in RNA-related assays and metabolic studies. It is supplied as a solid or as ready-to-use solutions for laboratory workflows and quality-control applications.

  • Ribonucleotide reagent for biochemical and cell-culture assays.
  • CAS 58-97-9; molecular formula C9H13N2O9P; molecular weight 324.18 g/mol.
  • Available as powder or prepared solutions (examples: 10 mM solution, 1 mL ampoules).
  • High purity reported (typically ≈99.8-99.98% in supplier documentation).
  • Soluble in water and DMSO; powder storage -20 °C, solutions typically -80 °C for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.