Learn More
Medchemexpress LLC Uridine 5'-monophosphate | 58-97-9 | MFCD00067346 | 99.8% | 324.18 g/mol | C9H13N2O9P | 50 MG
SDP
Catalog No. 5000247890
Encompass_Preferred
Uridine 5'-monophosphate (UMP) is a pyrimidine ribonucleotide used as a biochemical research reagent and metabolite reference. It is commonly used in enzymatic assays, nucleotide metabolism studies, and as a standard for analytical methods. The material is supplied in small pack sizes for laboratory use and is characterized for purity and identity.
  • Suitable as a metabolite reference and biochemical reagent.
  • High purity for reproducible analytical and enzymatic assays.
  • Provided in small pack sizes for research-scale experiments.
  • Water-soluble for straightforward aqueous formulations.
  • Characterized with molecular formula and molecular weight for identification.
Catalog No. 50-002-47890 Supplier Medchemexpress LLC Supplier No. HY101981R50MG
Uridine 5'-monophosphate (UMP) is a pyrimidine ribonucleotide used as a biochemical research reagent and metabolite reference. It is commonly used in enzymatic assays, nucleotide metabolism studies, and as a standard for analytical methods. The material is supplied in small pack sizes for laboratory use and is characterized for purity and identity.
  • Suitable as a metabolite reference and biochemical reagent.
  • High purity for reproducible analytical and enzymatic assays.
  • Provided in small pack sizes for research-scale experiments.
  • Water-soluble for straightforward aqueous formulations.
  • Characterized with molecular formula and molecular weight for identification.