Learn More
Medchemexpress LLC Uridine 5'-monophosphate-d11 dilithium | 00-00-0 | 99.9% | 347.12 g/mol | C9D11Li2N2O9P | 1 MG
SDP
Catalog No. 5000248045
Encompass_Preferred
Uridine 5'-monophosphate-d11 dilithium is a deuterium-labeled nucleotide provided as the dilithium salt for use as an internal standard or tracer in analytical and metabolic studies. It is intended for precise mass-spectrometry quantification and research applications; store frozen for stability.
  • Deuterium-labeled analogue for mass spectrometry tracing.
  • Dilithium salt form for improved handling and solubility.
  • High chemical purity (99.9%).
  • Supplied as a research-grade micro quantity (1 mg).
  • Stable when stored at -20°C for up to 2 years.
  • Suitable as an internal standard in LC-MS/MS workflows.
Catalog No. 50-002-48045 Supplier Medchemexpress LLC Supplier No. HY101981S21MG
Uridine 5'-monophosphate-d11 dilithium is a deuterium-labeled nucleotide provided as the dilithium salt for use as an internal standard or tracer in analytical and metabolic studies. It is intended for precise mass-spectrometry quantification and research applications; store frozen for stability.
  • Deuterium-labeled analogue for mass spectrometry tracing.
  • Dilithium salt form for improved handling and solubility.
  • High chemical purity (99.9%).
  • Supplied as a research-grade micro quantity (1 mg).
  • Stable when stored at -20°C for up to 2 years.
  • Suitable as an internal standard in LC-MS/MS workflows.