Learn More
Medchemexpress LLC Uridine 5'-monophosphate disodium salt | 3387-36-8 | 98.0% | 368.14 | 1 G
SDP
Catalog No. 5000454330
Encompass_Preferred
Uridine 5'-monophosphate disodium salt (Standard) is an analytical standard intended for research and analytical applications. It acts as an orally active mitochondrial ATP-dependent potassium channel activator, offering protective effects on the heart. This compound promotes CDP-choline synthesis and induces apoptosis in intestinal epithelial cells, supporting gut development and reducing diarrhea.
  • Analytical standard for research and analytical applications
  • Acts as an orally active mitochondrial ATP-dependent potassium channel activator
  • Offers protective effects on the heart
  • Promotes synthesis of CDP-choline
  • Induces apoptosis in intestinal epithelial cells
  • Beneficial for gut development
  • Reduces diarrhea
  • Used as a reference standard for assays
  • Utilized in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
Catalog No. 50-004-54330 Supplier Medchemexpress LLC Supplier No. HYW0131751G
Uridine 5'-monophosphate disodium salt (Standard) is an analytical standard intended for research and analytical applications. It acts as an orally active mitochondrial ATP-dependent potassium channel activator, offering protective effects on the heart. This compound promotes CDP-choline synthesis and induces apoptosis in intestinal epithelial cells, supporting gut development and reducing diarrhea.
  • Analytical standard for research and analytical applications
  • Acts as an orally active mitochondrial ATP-dependent potassium channel activator
  • Offers protective effects on the heart
  • Promotes synthesis of CDP-choline
  • Induces apoptosis in intestinal epithelial cells
  • Beneficial for gut development
  • Reduces diarrhea
  • Used as a reference standard for assays
  • Utilized in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS