Learn More
Medchemexpress LLC Uridine 5'-monophosphate disodium salt | 3387-36-8 | 99.9% | 368.14 | 5 G
SDP
Catalog No. 5000454414
Encompass_Preferred
Uridine 5'-monophosphate (5'-Uridylic acid) disodium salt is an orally active mitochondrial ATP-dependent potassium channel activator that has a protective effect on the heart. It can promote the synthesis of CDP-choline and induce apoptosis in intestinal epithelial cells, which is beneficial for gut development and reduces diarrhea.
  • Promotes late apoptosis in pig intestinal epithelial cells IPEC-J2.
  • Promotes apoptosis and renewal of intestinal villus tip epithelial cells in pigs.
  • Benefits gut development and and reduces diarrhea.
  • Enhances the synthesis of CDP-choline in the brains of gerbils.
  • Has protective properties for the heart in rats, preventing ischemic injury and some arrhythmias.
Catalog No. 50-004-54414 Supplier Medchemexpress LLC Supplier No. HYW0131755G
Uridine 5'-monophosphate (5'-Uridylic acid) disodium salt is an orally active mitochondrial ATP-dependent potassium channel activator that has a protective effect on the heart. It can promote the synthesis of CDP-choline and induce apoptosis in intestinal epithelial cells, which is beneficial for gut development and reduces diarrhea.
  • Promotes late apoptosis in pig intestinal epithelial cells IPEC-J2.
  • Promotes apoptosis and renewal of intestinal villus tip epithelial cells in pigs.
  • Benefits gut development and and reduces diarrhea.
  • Enhances the synthesis of CDP-choline in the brains of gerbils.
  • Has protective properties for the heart in rats, preventing ischemic injury and some arrhythmias.