Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Uridine 5'-(trihydrogen diphosphate), 4-oxime | 39023-73-9 | 98.8% | 419.18 | 10 MG
SDP

Catalog No. 5000365645
Encompass_Preferred

NHC-diphosphate is an active phosphorylated intracellular metabolite of β-d-N4-Hydroxycytidine (NHC) in a diphosphate form. NHC is a pyrimidine ribonucleoside and acts as a potent anti-virus agent, effectively inhibiting the replication of Venezuelan equine encephalitis virus (VEEV), Chikungunya virus (CHIKV), and hepatitis C virus (HCV).

  • Active phosphorylated intracellular metabolite
  • Potent anti-virus agent
  • Inhibits replication of Venezuelan equine encephalitis virus (VEEV)
  • Inhibits replication of Chikungunya virus (CHIKV)
  • Inhibits replication of hepatitis C virus (HCV)
  • For research use only
  • Soluble in H2O: 160 mg/mL (needs ultrasonic)
  • Store at -20°C, protect from light, stored under nitrogen

Catalog No. 50-003-65645 Supplier Medchemexpress LLC Supplier No. 10MGHY135867D
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NHC-diphosphate is an active phosphorylated intracellular metabolite of β-d-N4-Hydroxycytidine (NHC) in a diphosphate form. NHC is a pyrimidine ribonucleoside and acts as a potent anti-virus agent, effectively inhibiting the replication of Venezuelan equine encephalitis virus (VEEV), Chikungunya virus (CHIKV), and hepatitis C virus (HCV).

  • Active phosphorylated intracellular metabolite
  • Potent anti-virus agent
  • Inhibits replication of Venezuelan equine encephalitis virus (VEEV)
  • Inhibits replication of Chikungunya virus (CHIKV)
  • Inhibits replication of hepatitis C virus (HCV)
  • For research use only
  • Soluble in H2O: 160 mg/mL (needs ultrasonic)
  • Store at -20°C, protect from light, stored under nitrogen

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.