Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Uridine 5'-(trihydrogen diphosphate), sodium salt (1:x) | 21931-53-3 | 98.0% | 250 MG
SDP

Catalog No. 5000451575
Encompass_Preferred

Uridine 5'-diphosphate sodium salt is a potent, selective P2Y6 receptor native agonist and a potent P2Y14 antagonist. It is an endogenous metabolite involved in nucleic acid (RNA) biosynthesis and catalyzes the glucuronidation of various substrates. This product is for research use only.

  • Potent, selective P2Y6 receptor native agonist (EC50=300 nM; pEC50=6.52)
  • Potent P2Y14 antagonist (pEC50=7.28)
  • Induces microglial CCL2 and CCL3 mRNA expression
  • Induces chemokine expression in microglia
  • Activates NFATc1 and NFATc2 in microglia
  • White to off-white solid appearance
  • Store at -20°C under nitrogen

Catalog No. 50-004-51575 Supplier Medchemexpress LLC Supplier No. 250MGHYW010820
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Uridine 5'-diphosphate sodium salt is a potent, selective P2Y6 receptor native agonist and a potent P2Y14 antagonist. It is an endogenous metabolite involved in nucleic acid (RNA) biosynthesis and catalyzes the glucuronidation of various substrates. This product is for research use only.

  • Potent, selective P2Y6 receptor native agonist (EC50=300 nM; pEC50=6.52)
  • Potent P2Y14 antagonist (pEC50=7.28)
  • Induces microglial CCL2 and CCL3 mRNA expression
  • Induces chemokine expression in microglia
  • Activates NFATc1 and NFATc2 in microglia
  • White to off-white solid appearance
  • Store at -20°C under nitrogen

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.