Learn More
Medchemexpress LLC Uridine 5'-triphosphate-13C9 (dilithium) | 00-00-0 | 98.0% | 504.94 g/mol | 13C9H15N2O15P3·2Li | 1 MG
SDP
Catalog No. 5000241385
Encompass_Preferred
Uridine 5'-triphosphate-13C9 (dilithium) is an isotopically labeled nucleotide reagent used as a tracer or internal standard in biochemical and analytical workflows. Provided as the dilithium salt in aqueous Tris-HCl solution, it supports enzymatic assays, mass spectrometry, and metabolic labeling applications.
  • Isotopically labeled at nine carbons (13C9) for tracer and quantitation studies.
  • Dilithium salt form for enhanced solubility and stability in solution.
  • Provided as a concentrated solution suitable for direct use in assays.
  • High chemical purity for accurate analytical measurements.
  • Suitable for mass spectrometry, enzymology, and metabolic tracing.
Catalog No. 50-002-41385 Supplier Medchemexpress LLC Supplier No. HY107372S21MG
Uridine 5'-triphosphate-13C9 (dilithium) is an isotopically labeled nucleotide reagent used as a tracer or internal standard in biochemical and analytical workflows. Provided as the dilithium salt in aqueous Tris-HCl solution, it supports enzymatic assays, mass spectrometry, and metabolic labeling applications.
  • Isotopically labeled at nine carbons (13C9) for tracer and quantitation studies.
  • Dilithium salt form for enhanced solubility and stability in solution.
  • Provided as a concentrated solution suitable for direct use in assays.
  • High chemical purity for accurate analytical measurements.
  • Suitable for mass spectrometry, enzymology, and metabolic tracing.