Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Uridine 5'-triphosphate trisodium dihydrate | 116295-90-0 | MFCD00150714 | 97.0% | 586.12 | C9H16N2Na3O17P3 | 50 MG
SDP

Catalog No. 5000241529
Encompass_Preferred

Uridine 5'-triphosphate trisodium dihydrate is the trisodium salt hydrate of uridine 5'-triphosphate used as a biochemical reagent in molecular biology and pharmacology research. It serves as a nucleotide substrate and P2Y receptor agonist, and is used in enzymatic assays, nucleotide signaling studies, and RNA synthesis applications. The material is supplied as a high-purity, crystalline powder suitable for research use.

  • Purity 97.0% suitable for research assays.
  • Molecular weight 586.12 g/mol for formulation calculations.
  • Chemical formula C9H16N2Na3O17P3.
  • Supplied as a trisodium dihydrate salt, white to off-white powder.
  • Commonly used in nucleotide signaling, enzymology, and RNA synthesis studies.
  • Manufacturer datasheet, COA, and SDS available for quality and safety information.

Catalog No. 50-002-41529 Supplier Medchemexpress LLC Supplier No. HY107372A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Uridine 5'-triphosphate trisodium dihydrate is the trisodium salt hydrate of uridine 5'-triphosphate used as a biochemical reagent in molecular biology and pharmacology research. It serves as a nucleotide substrate and P2Y receptor agonist, and is used in enzymatic assays, nucleotide signaling studies, and RNA synthesis applications. The material is supplied as a high-purity, crystalline powder suitable for research use.

  • Purity 97.0% suitable for research assays.
  • Molecular weight 586.12 g/mol for formulation calculations.
  • Chemical formula C9H16N2Na3O17P3.
  • Supplied as a trisodium dihydrate salt, white to off-white powder.
  • Commonly used in nucleotide signaling, enzymology, and RNA synthesis studies.
  • Manufacturer datasheet, COA, and SDS available for quality and safety information.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.