Learn More
Abcam Uridine-5'-triphosphate [UTP] trisodium salt, P2Y receptor agonist, 100MG
Catalog No. 502704830
Encompass_Preferred
MW 550.09 g/mol, Purity >98%. Pyrimidine nucleoside triphosphate. Precursor in the enzymatic biosynthesis of RNA. P2Y receptor agonist, used to study ion transport functions of P2Y2 receptors on rat basolateral membranes.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4830 Supplier Abcam Supplier No. AB146222100MG
MW 550.09 g/mol, Purity >98%. Pyrimidine nucleoside triphosphate. Precursor in the enzymatic biosynthesis of RNA. P2Y receptor agonist, used to study ion transport functions of P2Y2 receptors on rat basolateral membranes.
The product is subject to the following: Abcam Restricted Use Statement