Learn More
Selleck Chemical LLC Uridine-50mg
SDP
Supplier:  Selleck Chemical LLC S202950MG
Encompass_Preferred
Uridine (NSC 20256,β-Uridine, Uridin) is a nucleoside, contains a uracil attached to a ribose ring via a β-N1-glycosidic bond.
Catalog No. 50-238-9830