Learn More
Apexbio Technology LLC Uridine, 50mg. Cas: 58-96-8 MFCD: MFCD00006526. DNA/RNA synthesis chemical
SDP
Catalog No. 501905243
Encompass_Preferred
Uridine, Trisodium Salt is an energy-rich precursor in the enzymatic biosynthesis of RNA, a potent vasodilator, and induces contractile responses in some tissues. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5243 Supplier Apexbio Technology LLC Supplier No. B147350
 Restricted Item
Add to Cart
Add to Cart
Uridine, Trisodium Salt is an energy-rich precursor in the enzymatic biosynthesis of RNA, a potent vasodilator, and induces contractile responses in some tissues. Other sizes are available. Please inqury us for quote.