Learn More
Medchemexpress LLC Uridine | 58-96-8 | 100.0% | 1 G
SDP
Catalog No. 5000413697
Encompass_Preferred
Uridine (β-Uridine) is a nucleoside compound consisting of uracil and a ribose ring, connected by a β-N1-glycosidic bond.
  • For research use only.
  • An endogenous metabolite.
  • Inhibits the proliferation of leukemia cell HL-60 and induces differentiation.
  • Alleviates CCl4-induced liver fibrosis and suppresses NF-κB signaling pathway.
  • Exhibits anti-ischemic, antioxidant, and anti-arrhythmic effects in rat models.
Catalog No. 50-004-13697 Supplier Medchemexpress LLC Supplier No. HYB14491G
Uridine (β-Uridine) is a nucleoside compound consisting of uracil and a ribose ring, connected by a β-N1-glycosidic bond.
  • For research use only.
  • An endogenous metabolite.
  • Inhibits the proliferation of leukemia cell HL-60 and induces differentiation.
  • Alleviates CCl4-induced liver fibrosis and suppresses NF-κB signaling pathway.
  • Exhibits anti-ischemic, antioxidant, and anti-arrhythmic effects in rat models.