Learn More
Medchemexpress LLC Uridine | 58-96-8 | 100.0% | 10 MM * 1 ML
SDP
Catalog No. 5000413394
Encompass_Preferred
Uridine (β-Uridine) is a nucleoside compound composed of uracil and a ribose ring, connected by a β-N1-glycosidic bond. This product is for research use only.
  • Inhibits proliferation of certain leukemia cells
  • Induces cell differentiation
  • Alleviates liver fibrosis
  • Inhibits pro-inflammatory cytokines
  • Activates the NF-κB signaling pathway
  • Provides anti-ischemic, antioxidant, and anti-arrhythmic effects
  • Protects against acute myocardial ischemia and reperfusion injury
Catalog No. 50-004-13394 Supplier Medchemexpress LLC Supplier No. HYB144910MM/1ML
Uridine (β-Uridine) is a nucleoside compound composed of uracil and a ribose ring, connected by a β-N1-glycosidic bond. This product is for research use only.
  • Inhibits proliferation of certain leukemia cells
  • Induces cell differentiation
  • Alleviates liver fibrosis
  • Inhibits pro-inflammatory cytokines
  • Activates the NF-κB signaling pathway
  • Provides anti-ischemic, antioxidant, and anti-arrhythmic effects
  • Protects against acute myocardial ischemia and reperfusion injury