Learn More
Medchemexpress LLC Uridine | 58-96-8 | 100.0% | 25 G
SDP
Catalog No. 5000414404
Encompass_Preferred
Uridine (β-Uridine) is a nucleoside compound composed of uracil and a ribose ring, which are connected by a β-N1-glycosidic bond. It is for research use only.
  • Inhibits proliferation of leukemia cell HL-60 and induces differentiation.
  • Alleviates CCl4-induced liver fibrosis by regulating cell activity and inhibiting pro-inflammatory cytokines.
  • Exhibits anti-ischemic, antioxidant, and anti-arrhythmic effects in rat models.
  • Weakens the expression of ACSL4 in murine lung tissues.
  • Antiviral activity against HIV1 3B infected in human H9 cells.
  • Anti-inflammatory activity in mouse RAW264.7 cells.
Catalog No. 50-004-14404 Supplier Medchemexpress LLC Supplier No. 25GHYB1449
Uridine (β-Uridine) is a nucleoside compound composed of uracil and a ribose ring, which are connected by a β-N1-glycosidic bond. It is for research use only.
  • Inhibits proliferation of leukemia cell HL-60 and induces differentiation.
  • Alleviates CCl4-induced liver fibrosis by regulating cell activity and inhibiting pro-inflammatory cytokines.
  • Exhibits anti-ischemic, antioxidant, and anti-arrhythmic effects in rat models.
  • Weakens the expression of ACSL4 in murine lung tissues.
  • Antiviral activity against HIV1 3B infected in human H9 cells.
  • Anti-inflammatory activity in mouse RAW264.7 cells.