Learn More
Medchemexpress LLC Uridine | 58-96-8 | 100.0% | 5 G
SDP
Catalog No. 5000413527
Encompass_Preferred
Uridine is a nucleoside compound composed of uracil and a ribose ring, connected by a β-N1-glycosidic bond. This compound is used for research purposes and has demonstrated various biological activities.
  • Inhibits the proliferation of HL-60 leukemia cells
  • Alleviates CCl4-induced liver fibrosis in mice
  • Shows anti-ischemic, antioxidant, and anti-arrhythmic effects in rat models
Catalog No. 50-004-13527 Supplier Medchemexpress LLC Supplier No. HYB14495G
Uridine is a nucleoside compound composed of uracil and a ribose ring, connected by a β-N1-glycosidic bond. This compound is used for research purposes and has demonstrated various biological activities.
  • Inhibits the proliferation of HL-60 leukemia cells
  • Alleviates CCl4-induced liver fibrosis in mice
  • Shows anti-ischemic, antioxidant, and anti-arrhythmic effects in rat models