Learn More
Medchemexpress LLC Uridine | 58-96-8 | MFCD00006526 | 100.0% | 10 G
SDP
Catalog No. 5000413351
Encompass_Preferred
Uridine is a nucleoside compound composed of uracil and a ribose ring, connected by a β-N1-glycosidic bond. It is for research use only and not sold to patients.
  • Inhibits proliferation and induces differentiation of leukemia cells.
  • Alleviates liver fibrosis.
  • Exhibits anti-ischemic, antioxidant, and anti-arrhythmic effects.
Catalog No. 50-004-13351 Supplier Medchemexpress LLC Supplier No. HYB144910G
Uridine is a nucleoside compound composed of uracil and a ribose ring, connected by a β-N1-glycosidic bond. It is for research use only and not sold to patients.
  • Inhibits proliferation and induces differentiation of leukemia cells.
  • Alleviates liver fibrosis.
  • Exhibits anti-ischemic, antioxidant, and anti-arrhythmic effects.