Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Uridine | 58-96-8 | MFCD00006526 | 100.0% | 100 MG
SDP

Catalog No. 5000413353
Encompass_Preferred

Uridine (Standard) is an analytical standard used for research and analytical applications. It functions as a reference standard supplied assay in various experiments, including HPLC, GC, and MS. This compound is an endogenous metabolite and also a disease marker for Nervous System Disorder.

  • Used as a reference standard supplied assay
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
  • Molecular weight: 244.20
  • Formula: C9H12N2O6
  • Appearance: white to off-white solid
  • Endogenous metabolite and disease marker for Nervous System Disorder

Catalog No. 50-004-13353 Supplier Medchemexpress LLC Supplier No. HYB1449R100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Uridine (Standard) is an analytical standard used for research and analytical applications. It functions as a reference standard supplied assay in various experiments, including HPLC, GC, and MS. This compound is an endogenous metabolite and also a disease marker for Nervous System Disorder.

  • Used as a reference standard supplied assay
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
  • Molecular weight: 244.20
  • Formula: C9H12N2O6
  • Appearance: white to off-white solid
  • Endogenous metabolite and disease marker for Nervous System Disorder

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.