Learn More
Medchemexpress LLC Uridine-d2 | 82740-98-5 | 99.0% | 1 MG
SDP
Catalog No. 5000412698
Encompass_Preferred
Uridine-d2 (β-Uridine-d2) is the deuterium labeled Uridine. It can be used as a tracer or an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has gained attention due to its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-12698 Supplier Medchemexpress LLC Supplier No. HYB1449S51MG
Uridine-d2 (β-Uridine-d2) is the deuterium labeled Uridine. It can be used as a tracer or an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has gained attention due to its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.