Learn More
Medchemexpress LLC Uridine-d2 | 82740-98-5 | 99.0% | 5 MG
SDP
Catalog No. 5000412674
Encompass_Preferred
Uridine-d2 is the deuterium labeled Uridine. It is for research use only and can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has gained attention due to its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis
  • Suitable for NMR, GC-MS, or LC-MS applications
  • Deuteration may affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-12674 Supplier Medchemexpress LLC Supplier No. HYB1449S55MG
Uridine-d2 is the deuterium labeled Uridine. It is for research use only and can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has gained attention due to its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis
  • Suitable for NMR, GC-MS, or LC-MS applications
  • Deuteration may affect pharmacokinetic and metabolic profiles of drugs