Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Uridine diphosphate glucuronic acid | 2616-64-0 | 98.0% | 580.29 | C15H22N2O18P2 | 5MG
SDP

Supplier:  Medchemexpress LLC HY1259545MG

Encompass_Preferred

Uridine diphosphate glucuronic acid is a nucleotide sugar used as a cofactor and substrate in glycosylation and glucuronidation reactions. It is supplied as a research-grade solid at high purity for enzymology and biochemical assays, and is handled and stored at low temperatures to preserve activity.

  • High purity (98.0%).
  • Suitable as an enzymatic substrate or cofactor in glycosylation and glucuronidation studies.
  • Solid, white to off-white appearance for easy handling.
  • Molecular weight 580.29 g/mol and formula C15H22N2O18P2.
  • Available in multiple small pack sizes for research use.
  • Shipped with dry ice and recommended storage at -80°C.

Catalog No. 50-002-38045


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.