Learn More
Medchemexpress LLC Uridine diphosphate glucuronic acid | 2616-64-0 | 98.0% | 580.29 g/mol | C15H22N2O18P2 | 10MG
SDP
Catalog No. 5000235340
Encompass_Preferred
Uridine diphosphate glucuronic acid (UDP-GlcA) is a sugar nucleotide cofactor used in biochemical and enzymatic research. It participates in glucuronidation and nucleotide sugar metabolism and serves as a substrate for glycosyltransferases and UDP-glucose dehydrogenase assays. The reagent is supplied for research use in biochemical assays and pathway studies.
  • High purity (98.0%) for reliable assay performance.
  • Molecular weight 580.29 g/mol and formula C15H22N2O18P2 for precise calculations.
  • Available in small pack sizes including 10 mg for screening and method development.
  • Suitable as a substrate in enzymatic and glycosylation studies.
  • Provided with batch-specific COA and SDS for traceability.
Catalog No. 50-002-35340 Supplier Medchemexpress LLC Supplier No. HY12595410MG
Uridine diphosphate glucuronic acid (UDP-GlcA) is a sugar nucleotide cofactor used in biochemical and enzymatic research. It participates in glucuronidation and nucleotide sugar metabolism and serves as a substrate for glycosyltransferases and UDP-glucose dehydrogenase assays. The reagent is supplied for research use in biochemical assays and pathway studies.
  • High purity (98.0%) for reliable assay performance.
  • Molecular weight 580.29 g/mol and formula C15H22N2O18P2 for precise calculations.
  • Available in small pack sizes including 10 mg for screening and method development.
  • Suitable as a substrate in enzymatic and glycosylation studies.
  • Provided with batch-specific COA and SDS for traceability.