Learn More
Medchemexpress LLC Uridine diphosphate glucuronic acid | 2616-64-0 | 98.0% | 580.29 g/mol | C15H22N2O18P2 | 1MG
SDP
Catalog No. 5000239166
Encompass_Preferred
Uridine diphosphate glucuronic acid (UDP-α-D-glucuronic acid) is a nucleotide sugar cofactor used as the glucuronic acid donor in enzymatic glucuronidation and polysaccharide biosynthesis studies. Supplied as a high-purity research reagent, the product is provided in milligram pack sizes and includes supporting documentation for quality and safe handling.
  • High purity (98.0%) suitable for biochemical assays.
  • Provided with certificate of analysis and safety data sheet for verification.
  • Available in multiple milligram pack sizes to support small-scale experiments.
  • Used as a glucuronic acid donor in enzymatic and metabolic studies.
  • Stable when stored and handled according to provided instructions.
Catalog No. 50-002-39166 Supplier Medchemexpress LLC Supplier No. HY1259541MG
Uridine diphosphate glucuronic acid (UDP-α-D-glucuronic acid) is a nucleotide sugar cofactor used as the glucuronic acid donor in enzymatic glucuronidation and polysaccharide biosynthesis studies. Supplied as a high-purity research reagent, the product is provided in milligram pack sizes and includes supporting documentation for quality and safe handling.
  • High purity (98.0%) suitable for biochemical assays.
  • Provided with certificate of analysis and safety data sheet for verification.
  • Available in multiple milligram pack sizes to support small-scale experiments.
  • Used as a glucuronic acid donor in enzymatic and metabolic studies.
  • Stable when stored and handled according to provided instructions.