Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Uridine diphosphate glucuronic acid | 2616-64-0 | 98.0% | 580.29 g·mol⁻¹ | C15H22N2O18P2 | 25MG
SDP

Catalog No. 5000238169
Encompass_Preferred

Uridine diphosphate glucuronic acid is a nucleotide sugar that functions as the glucuronic acid donor in glucuronidation and polysaccharide biosynthesis. Supplied as a research-grade biochemical with documented purity and analytical documentation, it is used in enzymology, metabolic assays, and in vitro glycosylation studies.

  • Purity 98.0%.
  • Molecular weight 580.29 g·mol⁻¹; formula C15H22N2O18P2.
  • Used as a substrate or cofactor in glucuronyltransferase and enzymatic assays.
  • Suitable for in vitro metabolism and glycosylation studies.
  • Available in small research pack sizes including 25 mg.
  • Supplied with a certificate of analysis and safety data sheet.

Catalog No. 50-002-38169 Supplier Medchemexpress LLC Supplier No. HY12595425MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Uridine diphosphate glucuronic acid is a nucleotide sugar that functions as the glucuronic acid donor in glucuronidation and polysaccharide biosynthesis. Supplied as a research-grade biochemical with documented purity and analytical documentation, it is used in enzymology, metabolic assays, and in vitro glycosylation studies.

  • Purity 98.0%.
  • Molecular weight 580.29 g·mol⁻¹; formula C15H22N2O18P2.
  • Used as a substrate or cofactor in glucuronyltransferase and enzymatic assays.
  • Suitable for in vitro metabolism and glycosylation studies.
  • Available in small research pack sizes including 25 mg.
  • Supplied with a certificate of analysis and safety data sheet.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.