Learn More
Medchemexpress LLC Uridine diphosphate glucuronic acid | 2616-64-0 | MFCD15145135 | 98.0% | 580.29 g/mol | C15H22N2O18P2 | 50MG
SDP
Catalog No. 5000237812
Encompass_Preferred
Uridine diphosphate glucuronic acid (UDP-α-D-glucuronic acid) is a nucleotide sugar that donates glucuronic acid in enzymatic conjugation reactions. Supplied as a high-purity solid, it is used as a cofactor in in vitro assays of glucuronyltransferases and in studies of UDP-dependent metabolism. The compound has CAS 2616-64-0 and a molecular weight of 580.29 g/mol.
  • High purity (98.0%) solid suitable for biochemical assays.
  • Serves as a glucuronic acid donor for glucuronyltransferases.
  • Used in enzymatic and metabolic pathway studies.
  • Available in small research quantities for laboratory use.
  • Molecular formula C15H22N2O18P2 and molecular weight 580.29 g/mol.
Catalog No. 50-002-37812 Supplier Medchemexpress LLC Supplier No. HY12595450MG
Uridine diphosphate glucuronic acid (UDP-α-D-glucuronic acid) is a nucleotide sugar that donates glucuronic acid in enzymatic conjugation reactions. Supplied as a high-purity solid, it is used as a cofactor in in vitro assays of glucuronyltransferases and in studies of UDP-dependent metabolism. The compound has CAS 2616-64-0 and a molecular weight of 580.29 g/mol.
  • High purity (98.0%) solid suitable for biochemical assays.
  • Serves as a glucuronic acid donor for glucuronyltransferases.
  • Used in enzymatic and metabolic pathway studies.
  • Available in small research quantities for laboratory use.
  • Molecular formula C15H22N2O18P2 and molecular weight 580.29 g/mol.