Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Uridine (standard) | 58-96-8 | 100.0% | 244.20 g/mol | C9H12N2O6 | 10 MG
SDP

Catalog No. 5000298950
Encompass_Preferred

Uridine (Standard) is an analytical-grade uridine supplied as a solid for research and analytical use. It provides a reliable reference material for assay development, calibration, and quality control in biochemical and analytical workflows.

  • Analytical standard for uridine.
  • High purity (99.98%).
  • White to off-white solid form.
  • Molecular weight 244.20 g/mol.
  • Compact 10 mg package size for analytical applications.
  • Suitable for research and analytical workflows including method validation.

Catalog No. 50-002-98950 Supplier Medchemexpress LLC Supplier No. HYB1449R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Uridine (Standard) is an analytical-grade uridine supplied as a solid for research and analytical use. It provides a reliable reference material for assay development, calibration, and quality control in biochemical and analytical workflows.

  • Analytical standard for uridine.
  • High purity (99.98%).
  • White to off-white solid form.
  • Molecular weight 244.20 g/mol.
  • Compact 10 mg package size for analytical applications.
  • Suitable for research and analytical workflows including method validation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.