Learn More
Selleck Chemical LLC uridine triacetate
SDP
Catalog No. 5000003069
Encompass_Preferred
Uridine triacetate (Vistonuridine PN401) is an orally active prodrug of the naturally occurring nucleoside uridine It is used for the treatment of hereditary orotic aciduria (Xuriden) or for the emergency treatment of fluorouracil or capecitabine overdose or toxicity
Catalog No. 50-000-03069 Supplier Selleck Chemical LLC Supplier No. S648425MG
Uridine triacetate (Vistonuridine PN401) is an orally active prodrug of the naturally occurring nucleoside uridine It is used for the treatment of hereditary orotic aciduria (Xuriden) or for the emergency treatment of fluorouracil or capecitabine overdose or toxicity