Learn More
Apexbio Technology LLC Uridine triacetate 4105-38-8 100mg
SDP
Catalog No. 5000090278
Encompass_Preferred
Uridine triacetate is an orally bioavailable prodrug of uridine It is designed to undergo rapid intestinal absorption followed by enzymatic deacetylation in circulation thereby releasing free uridine Uridine triacetate exerts its biological activity primarily through metabolic conversion to uridine which supports studies of systemic toxicity induced by chemotherapeutic agents such as 5-fluorouracil (5-FU) and capecitabine Based on these pharmacological properties uridine triacetate holds research potential in examining mechanisms underlying chemotherapy-induced cardiac and central nervous system toxicity as well as in studying metabolic events related to uridine homeostasis and therapeutic intervention following drug overdose or excessive chemotherapeutic exposure
Catalog No. 50-000-90278 Supplier Apexbio Technology LLC Supplier No. C7150100
Uridine triacetate is an orally bioavailable prodrug of uridine It is designed to undergo rapid intestinal absorption followed by enzymatic deacetylation in circulation thereby releasing free uridine Uridine triacetate exerts its biological activity primarily through metabolic conversion to uridine which supports studies of systemic toxicity induced by chemotherapeutic agents such as 5-fluorouracil (5-FU) and capecitabine Based on these pharmacological properties uridine triacetate holds research potential in examining mechanisms underlying chemotherapy-induced cardiac and central nervous system toxicity as well as in studying metabolic events related to uridine homeostasis and therapeutic intervention following drug overdose or excessive chemotherapeutic exposure